Order Entry
United States
ContactUsLinkComponent
Quantofix Ammonium Test Kit, CTL Scientific Supply
Quantofix Ammonium Test Kit, CTL Scientific Supply
Catalog # 10218-504
Supplier:  Macherey-Nagel
CAS Number:  
undefined
Quantofix Ammonium Test Kit, CTL Scientific Supply
Catalog # 10218-504
Supplier:  Macherey-Nagel
Supplier Number:  91315
CAS Number:  

Specifications

  • Description:
    Ammonium Test Kit
  • Storage:
    RT
  • Cat. No.:
    10218-504
  • Range:
    0-10-25-50-100-200-400 mg/L NH4+
  • Supplier no.:
    91315

Specifications

About this item

Quantofix Ammonium test strips and reagent for the semi-quantitative determination of ammonium in solutions.

Quantofix Ammonium has a gradation of scale: 0-10-25-50-100-200-400 mg/L NH4+ yellow to orange.

Ordering information: Packaged as a box of 100, strips and reagent.