Polarizing Film Slide Mount
470164-712
- Description:Polarizing Film, Slide Mounted
- Cat. No.:470164-712
- Supplier no.:PLF020
A must for your Optics lab.
- Sturdy slide mounts
Polarizing film, mounted in 2×2" slide mounts. (Pack of 10)
: White plastic slide holders are available separately as an accessory.