GeneMate Individual PCR Tubes, with Frosted-Flat Cap, 0.2 ml, Scientific Specialties
Supplier: GeneMate
Certificates
About this item
Secure evaporation-resistant seal between cap lid and tube.
- Ultra-thin wall for optimum heat transfer and higher yield
- Crystal clear clarity allows easy content viewing for each well
Available in full range of colors.
Certified free from DNase, RNase, and detectable DNA.
Frequently Bought Together