Anti-MAPK4 Rabbit Polyclonal Antibody
Catalog # 80084-034
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Mitogen-Activated Protein Kinase 4
- Antigen Symbol:MAPK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal (542–557 aa)
- Cross Adsorption:No
- Antigen Synonyms:ERK-4|ERK4|PRKM4|p63-MAPK|p63MAPK
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80084-034
- Supplier no.:AP7503A
Specifications
About this item
Type: Primary
Antigen: MAPK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (542–557 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse