Anti-USP13 Rabbit Polyclonal Antibody
80083-666
: Abgent
- Antibody Type:Primary
- Antigen Name:Ubiquitin specific peptidase 13
- Antigen Symbol:USP13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal ( 196–211 aa)
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80083-666
- Supplier no.:AP2141A
Type: Primary
Antigen: USP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 196–211 aa)
Host: Rabbit
Isotype:
Reactivity: Human