Anti-UBE2N Rabbit Polyclonal Antibody
Catalog # 80083-600
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Ubiquitin-conjugating Enzyme E2N
- Antigen Symbol:UBE2N
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal ( 1–50 aa)
- Cross Adsorption:No
- Antigen Synonyms:Ubiquitin-conjugating enzyme E2 N|UBC13|BLU|UbcH-ben|Bendless-like ubiquitin-conjugating enzyme|UbcH13|HEL-S-71
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80083-600
- Supplier no.:AP2125A
Specifications
About this item
Type: Primary
Antigen: UBE2N
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal ( 1–50 aa)
Host: Rabbit
Isotype:
Reactivity: Human