Anti-DNMT3L (DNA (cytosine-5-)-methyltransferase 3-like) Rabbit Polyclonal Antibody
Catalog # 80087-756
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:DNA (cytosine-5-)-methyltransferase 3-like
- Antigen Symbol:DNMT3L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western Blot:Yes
- Size:400 µL
- Cross Adsorption:No
- Antigen Synonyms:OCAB|OBF1|BOB1
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80087-756
- Supplier no.:AP1062A
Specifications
About this item
Type: Primary
Antigen: DNMT3L (DNA (cytosine-5-)-methyltransferase 3-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse