Anti-HA tag Mouse Monoclonal Antibody
Catalog # 80087-652
Supplier: Abgent
Specifications
- Antibody Type:Tag
- Antigen Name:HA Tag
- Antigen Symbol:HA tag (HA tag)
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Western Blot:Yes
- Size:400 µL
- Cross Adsorption:No
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:80087-652
- Supplier no.:AM1008A
Specifications
About this item
Type: Tag
Antigen: HA tag (HA tag)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: