About this item
As an ISO Guide 34 accredited manufacturer, Microbiologics offers Lab-Elite Certified Reference Materials to help ISO 17025 accredited laboratories meet the requirements of this standard
- Traceable to reference culture
- Highly characterized microorganisms
- Supported by the highest accreditations in the industry
- For use in ISO 17025 accredited labs
- Quality Control of research
- For validation and verification of processes and instruments
Each kit Includes one self-contained device including a lyophilized QC microorganism pellet, reservoir of hydrating fluid, inoculating swab and a Certificate of Analysis in the canister.
Lab-Elite Certified Reference Material is delivered in an easy-to-use format and includes a Certificate of Analysis inside the canister. The Lab-Elite Certified Reference Material product line continues to expand with the addition of new microorganism strains in this convenient format.
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Related Information
Certificates
Search by Lot NumberInstructions for End User Registration with Microbiologics