Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Desmoglein-3
- Antigen Symbol:DSG3
- Clonality:Monoclonal
- Clone:5H10
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:1830
- Antigen Synonyms:Cadherin family member 6|desmoglein 3|desmoglein-3|PVA|desmoglein 3 (pemphigus vulgaris antigen)|CDHF6130-kD pemphigus vulgaris antigen|pemphigus vulgaris antigen|DKFZp686P23184,130 kDa pemphigus vulgaris antigen
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:The extracellular domain of human Desmoglein 3 [Swiss-Prot# P32926]
- Purification:Protein G purified
- Cat. No.:102714-548
- Supplier no.:NBP1-78984B
Specifications
About this item
The Desmoglein-3 Antibody (5H10) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to Desmoglein-3. This antibody reacts with human. The Desmoglein-3 Antibody (5H10) [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Desmoglein-3
Clonality: Monoclonal
Clone: 5H10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse