To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Pregnancy specific beta-1-glycoprotein 4
- Antigen Symbol:BMP-2
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:650
- Antigen Synonyms:BMP2ABone morphogenetic protein 2A|BMP-2|bone morphogenetic protein 2|BMP-2A
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:A synthetic peptide made to an internal region of human BMP2 (within residues 250-350) [Swiss-Prot# P12643]
- Purification:Immunogen affinity purified
- Cat. No.:102714-330
- Supplier no.:NBP1-19751G
The BMP-2 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to BMP-2. This antibody reacts with human, mouse, rat. The BMP-2 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: BMP-2
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat