Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphodiesterase 9A
- Antigen Symbol:PDE9A
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.05 ml
- Gene ID:5152
- Antigen Synonyms:FLJ90181|high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A|HSPDE9A2|CGMP-specific 3'-5'-cyclic phosphodiesterase type 9|phosphodiesterase 9A|EC 3.1.4.35|EC 3.1.4.1710phosphodiesterase PDE9A21
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:Sequence from the C-terminal region of PDE9A
- Purification:Immunogen affinity purified
- Cat. No.:102714-984
- Supplier no.:NBP1-00641B
Specifications
About this item
The PDE9A Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to PDE9A. This antibody reacts with human, mouse, rat. The PDE9A Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: PDE9A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat