Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody Type:Primary
- Antigen Name:Histone H3
- Antigen Symbol:HIST2H3C
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:C. elegans,Human,Mouse
- Western Blot:Yes
- Size:0.05 ml
- Gene ID:126961
- Antigen Synonyms:H3.3B|family 3A|MGC87782|H3.3AH3F3H3F3B|histone H3.3|MGC87783|H3 histone
- Modification:Monomethyl Arg2
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:Synthetic monomethylated peptide surrounding Arginine 2 of human Histone H3 [Monomethyl Arg2][Swiss Prot Q71DI3].
- Purification:Immunogen affinity purified
- Cat. No.:102714-706
- Supplier no.:NB21-1001B
Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: