Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Name:Hepatic Sinusoidal Endothelial Cells
- Clonality:Monoclonal
- Clone:SE-1
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Antigen Synonyms:Hepatic Sinusoidal Endothelial Cells|SE1|Sinusoidal Endothelial Cells|Hepatic cells|HSE
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:Rat Hepatic Sinusoidal Endothelial Cells
- Purification:Protein A purified
- Cat. No.:102713-972
- Supplier no.:NB110-68095B
Specifications
About this item
Type: Primary
Antigen: Hepatic Sinusoidal Endothelial Cells
Clonality: monoclonal
Clone: SE-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse, Rat