Order Entry
United States
ContactUsLinkComponent
 
Anti-GFP Tag Rabbit Polyclonal Antibody
  89495-082
 :  Genscript
undefined
Anti-GFP Tag Rabbit Polyclonal Antibody
  89495-082
 :  Genscript
 :  A01704-100

 

  • Antibody Type:
    Tag
  • Antigen Name:
    GFP Tag
  • Antigen Symbol:
    GFP Tag
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Antigen Synonyms:
    GFPuv|eGFP|Green Fluorescent Protein
  • Storage Buffer:
    PBS, pH 7.4, containing 0.02% sodium azide
  • Storage Temperature:
    The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
  • Shipping Temperature:
    Ice
  • Immunogen:
    purified recombinant full-length GFP protein
  • Purification:
    Affinity chromatography
  • Cat. No.:
    89495-082
  • Packaging:
    Vial
  • Supplier no.:
    A01704-100

 

 

GenScript THE™ GFP antibody, pAb, Rabbit specifically reacts with fusion proteins containing GFP epitope tag. This antibody can recognize all variants of GFP protein such as eGFP, AcGFP1 and cGFP. It slightly recognizes a variety of variants of RFP protein such as DsRed, DsRed2, mOrange, mCherry and tdTomato.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: