Order Entry
United States
ContactUsLinkComponent
Anti-c-Myc Tag Mouse Monoclonal Antibody [clone: 2G8D5] (Biotin)
Anti-c-Myc Tag Mouse Monoclonal Antibody [clone: 2G8D5] (Biotin)
Catalog # 89493-636
Supplier:  Genscript
undefined
Anti-c-Myc Tag Mouse Monoclonal Antibody [clone: 2G8D5] (Biotin)
Catalog # 89493-636
Supplier:  Genscript
Supplier Number:  A00864-200

Specifications

  • Antibody Type:
    Tag
  • Antigen Name:
    c-Myc Tag
  • Antigen Symbol:
    c-Myc Tag
  • Clonality:
    Monoclonal
  • Clone:
    2G8D5
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Western Blot:
    Yes
  • Size:
    200 μg
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Antigen Synonyms:
    bHLHe39|Myc2|Nird|AU016757|Niard
  • Storage Buffer:
    PBS, pH 7.4, containing 1% BSA and 0.02% sodium azide
  • Storage Temperature:
    The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/ml
  • Shipping Temperature:
    Ice
  • Immunogen:
    A synthetic peptide EQKLISEEDL coupled to KLH
  • Purification:
    Protein A affinity column
  • Cat. No.:
    89493-636
  • Packaging:
    Vial
  • Supplier no.:
    A00864-200

Specifications

About this item

GenScript THE™ c-Myc antibody [Biotin], mAb, Mouse recognizes C-terminal, N-terminal and internal c-Myc tagged fusion proteins expressed in prokaryotic or eukaryotic cells. It also recognizes c-Myc gene in human cancers, such as Hela whole cell lysates.

  • Comprehensive pool
  • Against 24 tags
  • Available in gram level bulk quantities
  • Various conjugates for hot tags detection

GenScript offers more than 90 highly specific monoclonal and polyclonal antibodies against over 30 different tags. They are pre-validated for a variety of immunological applications, such as western blot analysis, immunoprecipitation, immunofluorescence, and affinity purification. Our epitope tag antibodies are manufactured under extensive quality control to ensure optimal sensitivity and consistent high performance.

Type: Tag
Antigen: MYC
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: