About this item
EndoFree Plasmid Kits provide rapid, anion-exchange-based endotoxin-free plasmid DNA purification.
- Less than 0.1 EU/µg DNA achieved
- Fast procedure, with integrated endotoxin removal
- High yields of high-copy plasmid DNA
- LyseBlue for optimum lysis and maximum DNA yield
Kit enables the purification of up to 10 mg endotoxin free advanced transfection grade plasmid or cosmid DNA. QIAfilter Cartridges enable fast lysate clearing by filtration. The purified DNA exceeds the purity obtained by 2 x CsCl gradient centrifugation and is suitable for advanced transfection-grade applications. The EndoFree Plasmid Buffer Set can be used for preparations of 10 mega or 5 giga transfection-grade plasmid or cosmid DNA preparations.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: