ContactUsLinkComponent
ESD Safe Sheet Protectors, Transforming Technologies
ESD Safe Sheet Protectors, Transforming Technologies
Catalog # 14224-700
undefined
ESD Safe Sheet Protectors, Transforming Technologies
Catalog # 14224-700
Supplier Number:  DC1185
Specifications
  • Color:
    Clear
  • Description:
    ESD Safe Sheet Protectors
  • Cat. No.:
    14224-700
  • Supplier no.:
    DC1185
Specifications
About this item
ESD sheet protectors or ESD safe document holders are designed for use in ESD controlled areas where electronic sensitive material is produced
  • Lightly embossed for glare reduction
  • Resistant to print transfer
  • White stripe with ESD identification
  • 3-hole punch
  • Surface resistivity of 10^9 - 10^11 @ 15% RH. Std.
All sheet protectors are made from 2 mil, stable, amine-free, polypropylene clear film, and the material is specially treated to reduce 90% of charge generation in less than 1 second per NFPA-56A and MIL B 81705B.