Wrist Strap and Coil Cord Set, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The wrist strap series 360° curved wrist band and coil cord set features hypoallergenic silver-plated nylon thread for conductivity
The band is standard with a 4 mm, 7 mm or 10 mm male snap or a 4 mm machined post and has a maximum resistance of 160 ohms. The soft, blue woven fabric and curved buckle of the WB0016 wrist band allows the skin to breathe and makes it very comfortable to wear. The coil cord’s 2.5 mm polyurethane coil insulation offers excellent coil memory and a swivel type banana jack increases coil life and prevents cord tangles.
Certifications: Meets or exceeds requirements of ANSI ESDS20.20 and ESDA Standard 1.1-2006
Frequently Bought Together