Woven Wrist Band, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
The soft, blue woven fabric and curved buckle of the woven wrist band allows the skin to breathe and makes it very comfortable to wear
- Polyester fabric with silver-plated nylon
- 100 ohms typical resistance
- 360 degrees contact around wrist
- Blue in color with a stainless steel back plate
- 1 to 5 pound snap release
- Available in 4 mm, 7 mm, 10mm, or 4 mm machined post
These wristbands use hypoallergenic silver-plated nylon thread for conductivity and has a maximum resistance of 160 ohms.
Certifications: Meets or exceeds requirements of ANSI ESDS20.20 and ESDA Standard 1.1-2006
Frequently Bought Together