Specifications
- Description:Drug Screen Urine Test Panel, iCassette®
- Size:25 drug tests/kit
- Cat. No.:89171-328
- Parameters tested:AMP, BAR, BZO, COC, MDMA, MOP, OXY, PCP, PPX, TCA, THC
- Supplier no.:I-DCB-1115-011
Specifications
About this item
Rapid Drug Screen for the simultaneous, qualitative detection of single or multiple drug metabolites in human urine
Pipette style drug screen uses three drops of urine per well to detect drug metabolites in urine, with results in 5 minutes.
Room temperature storage, long shelf-life. Pipette included. Collection cup not included.
Rapid screen for a collection of illicit and prescription drugs. Each kit contains materials for 25 drug tests. Rapid Drug Screens can include Amphetamine (AMP), Barbiturates (BAR), Benzodiazepines (BZO), Buprenorphine (BUP), Cocaine (COC), Methamphetamine (mAMP), Opiates (OPI) and (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Propoxyphene (PPX), Ecstacy (MDMA), Tricyclic Antidepressents (TCA), and Marijuana (THC) in human urine at the following cutoff levels (ng/mL) unless otherwise noted in description: AMP (1000), BAR (300), BZO (300), BUP (10), COC (300), mAMP (1000), OPI (2000), MOP (300), MTD (300), OXY (100), PCP (25), PPX (300), MDMA (500), TCA (1000), and THC (50).