Order Entry
United States
Orders LinkContactUsLinkComponent
 

Sodium hydroxide 10 N

1 L
Catalog # 100217-766
Corrosive
Supplier:   Teknova
 :  N4710
Sodium hydroxide 10 N
Catalog # 100217-766
Supplier:  Teknova
CAS Number:  1310-73-2
Sodium hydroxide 10 N

1 L

CAS Number:  1310-73-2
Price
$147.33
Log in for availability
Each (1l)
 
Product Details & Documents

Sterile Sodium Hydroxide, store it in room temperature.

Synonyms:  Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • UN:1823
  • ADR:8,II
Specifications

Specifications

  • Size:
    1 L

Specifications