Specifications
- Description:R2A Broth premix
- Size:500 g
- Environmentally Preferable:
- Cat. No.:101454-176
- Supplier no.:R0005
Specifications
About this item
R2A Media is used for the growth of heterotrophic organisms found in potable water.
- Heterotrophic organism growth
- Meets the needs of molecular biologists
- High-quality components
Teknova's Dry Media is formulated to meet the needs of a molecular biologist. It made of the highest quality components and specifically tested for solubility, color, clarity,conductivity, pH, and growth assays. These convenient pre-mixes only require the addition of water and autoclaving. Use 3.15 g of premix per litre of water, and autoclave to sterilize.