Anti-PRPF6 Rabbit Polyclonal Antibody
Catalog # 10273-170
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Pre-mRNA Processing Factor 6
- Antigen Symbol:PRPF6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:150 µL
- Cross Adsorption:No
- Antigen Synonyms:S. cerevisiae|androgen receptor N-terminal domain transactivating|hPrp6|U5-102K|PRP6 pre-mRNA processing factor 6 homolog|C20orf14|TOM|ANT-1|U5 snRNP-associated 102 kDa protein
- Storage Buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage Temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity purified
- Cat. No.:10273-170
- Supplier no.:23929-1-AP
Specifications
About this item
The PRPF6 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human PRPF6. This antibody recognizes human, mouse antigen. The PRPF6 antibody has been validated for the following applications: ELISA, WB, IHC analysis.
Type: Primary
Antigen: PRPF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse