Anti-MSX2 Mouse Monoclonal Antibody [clone: 2E12]
Catalog # 10049-790
Supplier: Enzo Life Sciences
Specifications
- Antibody Type:Primary
- Antigen Name:Msh Homeobox 2
- Antigen Symbol:MSX2
- Clonality:Monoclonal
- Clone:2E12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Form:Liquid
- Antigen Synonyms:MSH|FPP|HOX8|PFM|PFM1|CRS2
- Storage Buffer:PBS, pH 7.2, containing 50% glycerol.
- Storage Temperature:–20 °C
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant human Msx2 (N-terminal fragment).
- Purification:purified by Protein A affinity chromatography
- Cat. No.:10049-790
- Supplier no.:ADI-AAM-010-E
Specifications
About this item
Msx2, also known as HOX-8, is a homeobox containing transcription factor, which is a member of the MSH family of homeobox proteins. Homeobox (HOX) genes encode DNA binding proteins that recognize specific sequences and modulate transcriptional activity. Msx2 is believed to play a role in regulating bone development, particularly in sutures of cranial bone.
Type: Primary
Antigen: MSX2
Clonality: Monoclonal
Clone: 2E12
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human