Anti-MTTF1 Rabbit Polyclonal Antibody
Catalog # 10209-620
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Transcription Factor A, mitochondrial
- Antigen Symbol:MTTF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:mitochondrial transcription factor A|mtTFA|TCF-6|TCF6L1|mitochondrial transcription factor 1|Transcription factor 6-like 2 (mitochondrial transcription factor)|TCF6L2|transcription factor 6|TCF6L3|TCF6|MtTF1
- Amino Acid Number:227 to 246
- Sequence:RKDLLRRTIKKQRKYGAEEC
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human mtTFA (227-246aa RKDLLRRTIKKQRKYGAEEC).
- Purification:purified by immunogen affinity purification
- Cat. No.:10209-620
- Supplier no.:PA1936
Specifications
About this item
Rabbit IgG polyclonal antibody for Transcription factor A, mitochondrial(TFAM) detection. Tested with WB, IHC-P, IHC-F, ICC in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: MTTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human