To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:NAPDH oxidase 1
- Antigen Symbol:NOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:NOH-1|MOX1|NOX-1|NOH1|GP91-2|NADPH oxidase 1|Mitogenic oxidase 1
- Amino Acid Number:354 to 374
- Sequence:HIRAAGDWTENLIRAFEQQYS
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human NOX1 (354-374aa, HIRAAGDWTENLIRAFEQQYS).
- Purification:purified by immunogen affinity purification
- Cat. No.:10207-256
- Supplier no.:PA1739
Specifications
About this item
Rabbit IgG polyclonal antibody for NADPH oxidase 1(NOX1) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: NOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human