Anti-ATF4 Rabbit Polyclonal Antibody
Catalog # 10207-176
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Activating Transcription Factor 4
- Antigen Symbol:ATF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:DNA-binding protein TAXREB67|cAMP-dependent transcription factor ATF-4|Cyclic AMP-dependent transcription factor ATF-4|TXREB|TaxREB67|Tax-responsive enhancer element-binding protein 67|CREB2|Activating transcription factor 4|cAMP-responsive element-binding protein 2|CREB-2|Cyclic AMP-responsive element-binding protein 2|ATF4
- Amino Acid Number:332 to 351
- Sequence:QYLKDLIEEVRKARGKKRVP
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ATF4 (332-351aa QYLKDLIEEVRKARGKKRVP), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. No.:10207-176
- Supplier no.:PA1537
Specifications
About this item
Rabbit IgG polyclonal antibody for Cyclic AMP-dependent transcription factor ATF-4(ATF4) detection. Tested with WB in Human;Mouse;Rat.
- Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: ATF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat