Anti-MAP2K3 Rabbit Polyclonal Antibody
100 µg
Catalog # 10207-158
Supplier: Boster Biological Technology
Supplier #: PA1377-1
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit IgG polyclonal antibody for Dual specificity mitogen-activated protein kinase kinase 3(MAP2K3) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: MAP2K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
Specifications
- Size:100 µg
- Cross Adsorption:No
- Clonality:Polyclonal
- Purification:purified by immunogen affinity purification
- Epitope:near the N-terminus
- Form:Lyophilized
- Isotype:IgG
- Host:Rabbit
- Sequence:RNLDSRTFITIGDRNFE
- Cat. No.:10207-158
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:MAP2K3
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human MEK3 (42-58aa RNLDSRTFITIGDRNFE), identical to the related mouse and rat sequences.
- Amino Acid Number:42 to 58
- Conjugation:Unconjugated
- Antigen Synonyms:SAPKK-2|MEK 3|MAP kinase kinase 3|MEK3|SAPK kinase 2|SAPKK2|PRKMK3|SKK2|MKK3|MAPKK 3|MAP2K3|Stress-activated protein kinase kinase 2|MAPK/ERK kinase 3|Dual specificity mitogen-activated protein kinase kinase 3
- Antigen Name:Dual specificity mitogen-activated protein kinase kinase 3
- Storage Temperature:–20 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



