Anti-ERK2 Rabbit Polyclonal Antibody
Catalog # 10207-122
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Extracellular signal-regulated kinase 2
- Antigen Symbol:ERK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:PRKM2|Extracellular signal-regulated kinase 2|MAPK2|ERK|ERK-2|Mitogen-activated protein
- Amino Acid Number:348 to 360
- Sequence:FEETARFQPGYRS
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human ERK2 (348-360aa FEETARFQPGYRS), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. No.:10207-122
- Supplier no.:PA1310
Specifications
About this item
Rabbit IgG polyclonal antibody for Mitogen-activated protein kinase 1(MAPK1) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: ERK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat