Anti-TLR10 Rabbit Polyclonal Antibody
Catalog # 10206-956
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:Toll-like Receptor 10
- Antigen Symbol:TLR10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:Toll-like receptor 10|CD290
- Amino Acid Number:88 to 102
- Sequence:DLKTFEFNKELRYLD
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human TLR10 (88-102aa DLKTFEFNKELRYLD).
- Purification:purified by immunogen affinity purification
- Cat. No.:10206-956
- Supplier no.:PA1958
Specifications
About this item
Rabbit IgG polyclonal antibody for Toll-like receptor 10(TLR10) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: TLR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human