Anti-Episialin, EMA Mouse Monoclonal Antibody [clone: EMA-39]
10206-228
- Antibody Type:Primary
- Antigen Name:Episialin, EMA
- Antigen Symbol:EMA
- Clonality:Monoclonal
- Clone:EMA-39
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage Temperature:–20 °C
- Immunogen:Human milk fat globule membranes.
- Cat. No.:10206-228
- Supplier no.:MA1039
Mouse IgG monoclonal antibody for Episialin, EMA, mucin 1, cell surface associated (MUC1) detection. Tested with IHC-P, IHC-F in Human. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: Episialin, EMA
Clonality: monoclonal
Clone: EMA-39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human