Anti-CD3 Mouse Monoclonal Antibody [clone: CA-3]
Catalog # 10206-188
Supplier: Boster Biological Technology
Specifications
- Antibody Type:Primary
- Antigen Name:CD3 Molecule
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:CA-3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage Temperature:–20 °C
- Immunogen:Human thymocytes followed by Sezary T cells.
- Cat. No.:10206-188
- Supplier no.:MA1015
Specifications
About this item
Mouse IgG monoclonal antibody for CD3, CD3e molecule, epsilon (CD3-TCR complex) (CD3E ) detection. Tested with IHC-F in Human. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: CA-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: