To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cathepsin D
- Antigen Symbol:CTSD
- Clonality:Monoclonal
- Clone:CP-D13
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen Synonyms:lysosomal aspartyl protease|HEL-S-130P|ceroid-lipofuscinosis|lysosomal aspartyl peptidase|CPSD|CLN10|neuronal 10
- Storage Temperature:–20 °C
- Immunogen:Human liver cathepsin D.
- Cat. No.:10205-960
- Supplier no.:MA1013
Specifications
About this item
Mouse IgG monoclonal antibody for Cathepsin D, cathepsin D (CTSD) detection. Tested with WB, IHC-P in Human. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: CTSD
Clonality: Monoclonal
Clone: CP-D13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: