Anti-BIN1 Mouse Monoclonal Antibody [clone: 2F11]
RL200301E63S
- Antibody Type:Primary
- Antigen Name:Bridging integrator 1
- Antigen Symbol:BIN1
- Clonality:Monoclonal
- Clone:2F11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:25 µL
- Antigen Synonyms:box dependant MYC interacting protein 1|myc box-dependent-interacting protein 1|box-dependent myc-interacting protein 1|amphiphysin II|AMPH2|AMPHL|SH3P9|amphiphysin-like protein
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Cat. No.:RL200301E63S
- Supplier no.:200-301-E63S
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 2F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human