TransIT-X2® Dynamic Delivery System, Mirus Bio
TransIT-X2® dynamic delivery system is an advanced, non-liposomal polymeric system that enables high efficiency transfection of many cell types, including primary cells.
- Peak performance in a broad range of cell types
- Suitable for multiple applications
- Efficient delivery of plasmid DNA, siRNA/miRNA and CRISPR/Cas9 components
TransIT-X2® dynamic delivery System is in 80% ethanol and should be stored tightly capped at −20 °C.
TransIT-X2 can be used for DNA delivery, siRNA or miRNA delivery or simultaneous co-transfection of DNA and siRNA or miRNA. Transfections with TransIT-X2 do not require medium changes and can be carried out in serum-containing medium.
: TransIT-X2® dynamic delivery system is available in 0.3, 0.75, 1.5 and 5×1.5 ml (5-pack) and 10×1.5 ml (10-pack) configurations.
- CRISPR/Cas 9:Yes
- DNA and siRNA:Yes
- DNA:Yes
- siRNA:Yes
Frequently Bought Together