Order Entry
United States
ContactUsLinkComponent
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride Pd 13.5-15.5%
Irritant
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride Pd 13.5-15.5%
Catalog # AA46503-03
CAS Number:  
Irritant
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride Pd 13.5-15.5%
Catalog # AA46503-03
Supplier Number:  046503-03
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1g
  • Cat. No.:
    AA46503-03
  • Supplier no.:
    46503-03

Specifications

About this item

 :
Formula:C₃₄H₂₈Cl₂P₂FePd
MDL Number:MFCD00015757
 :731.70 g/mol
Melting Pt:266…283 °C
EINECS:460-040-3
 :GHS07
Storage Temperature:Ambient
CAS Number:72287-26-4