ContactUsLinkComponent
Syringes with BD Luer-Lok® Tip, Autosampler, PerkinElmer
Syringes with BD Luer-Lok® Tip, Autosampler, PerkinElmer
Catalog # 10129-657
Supplier:  PerkinElmer
Syringes with BD Luer-Lok® Tip, Autosampler, PerkinElmer
Catalog # 10129-657
Supplier:  PerkinElmer
Supplier Number:  02542891
Specifications
  • Volume:
    3000 µl (3 ml)
  • Description:
    Syringe with Luer-Lok® Tip
  • Cat. No.:
    10129-657
  • Supplier no.:
    02542891
Specifications
About this item
Syringe with BD Luer-Lok® Tip is a autosampler syringe useful in 200 series autosampler.
  • Increased secure connection
  • Tapered plunger rod
  • Positive plunger rod stop
  • Bold graduation markings
BD™ syringes feature a clear barrel with bold scale markings, tapered plunger rod for ease of aspiration, positive plunger rod stop, and an added BD Luer-Lok™ thread for increased secure connection.