ContactUsLinkComponent
Gel Loading Tips, Phenix Research Products
Gel Loading Tips, Phenix Research Products
Catalog # 101100-682
Gel Loading Tips, Phenix Research Products
Catalog # 101100-682
Supplier Number:  1158Q46
Specifications
  • Volume:
    200 µl
  • Packed:
    96 Tips/Rack, 10 Racks/Pack
  • Cat. No.:
    101100-682
  • Sterile:
    +
  • Supplier no.:
    T-061BRS
Specifications
About this item
These are sterile, universal fit, wide bore tips.
Delivery: Packaged in hinged racks.