Human TNFSF13/APRIL PicoKine ELISA Kit, Boster
Catalog # 10205-270
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:TNFSF13/APRIL
- Size:96 well
- Immunogen:Standard: NSO,A105-L250
- Cat. No.:10205-270
- Supplier no.:EK0921
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Human TNFSF13/APRIL
Ordering information: Kits include 96-well plate precoated with anti- human TNFSF13/APRIL antibody 1; ST0000-10 Lyophilized recombinant human TNFSF13/APRIL standard 10ng/tubex2; AR1107 Biotinylated anti- human TNFSF13/APRIL antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.