Human IL-8 PicoKine ELISA Kit, Boster
10205-740
:
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:IL-8 (CXCL8)
- Size:96 well
- Immunogen:Standard: E.coli, S28-S99
- Cat. No.:10205-740
- Supplier no.:EK0413
Sandwich ELISA kit of Quantitative Detection for Human IL-8
: Kits include 96-well plate precoated with anti- human IL-8 antibody 1; ST0000-10 Lyophilized recombinant human IL-8 standard 10ng/tubex2; AR1107 Biotinylated anti- human IL-8 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.