Order Entry
United States
ContactUsLinkComponent
Motorized Pipette Filler, Chemglass
Motorized Pipette Filler, Chemglass
Catalog # 89167-164
Supplier:  Chemglass
Small Business Enterprise
undefined
Motorized Pipette Filler, Chemglass
Catalog # 89167-164
Supplier:  Chemglass
Supplier Number:  CLS-3472-10
Small Business Enterprise

Specifications

  • Description:
    Motorized Pipette Filler
  • Compatible With:
    Glass or Plastic Pipettes (0.1- 100 mL), Pasture Pipettes
  • Dispensing Speed:
    8+ Gravity Dispense
  • Filter Type:
    Hydrophobic
  • Battery Type:
    Rechargeable Lithium-Ion
  • Cat. No.:
    89167-164
  • Supplier no.:
    CLS-3472-10

Specifications

About this item

Motorized pipette filler features a lightweight, ergonomic design allowing longer, fatigue-free pipetting.

  • Separate aspirate and dispense speed controls
  • Lithium-ion battery
  • Autoclavable nose cone housing

Features a full volume range of 0.1 to 100 ml with easy one-handed operation. A large LCD display provides visual confirmation of remaining battery charge and speed settings. The lithium-ion battery offers long run-time per charge without battery memory problems. The fully autoclavable nose cone housing simplifies disinfecting and decontamination.