Order Entry
United States
Orders LinkContactUsLinkComponent
ICT Accutase Cell Detachment Solution

ICT Accutase Cell Detachment Solution

100 ml
Catalog # 490007-741
Supplier #:  AT104
ICT Accutase Cell Detachment Solution
Catalog # 490007-741
CAS Number:  
ICT Accutase Cell Detachment Solution
ICT Accutase Cell Detachment Solution

100 ml

CAS Number:  
Price
$67.28
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Accutase can be used whenever gentle and efficient detachment of any adherent cell line is needed

Accutase performs exceptionally well in detaching cells for hESC culturing, analysis of cell surface markers, virus growth assay, quiescence assays by serum starvation, transformation assays by oncogene transfection, neural crest cell migration assays, cell proliferation, apoptosis, cell haptotaxsis, tumor cell migration assays, routine cell passage, production scale-up (bioreactor), and flow cytometry.

Accutase has been shown to detach a few cell lines without harm from hESCs, fibroblasts, keratinocytes, vascular endothelial cells, hepatocytes, vascular smooth muscle cells, hepatocyte progenitors, primary chick embryo neuronal cells, bone marrow stem cells, adherent CHO and BHK cells, macrophages, 293 cells, L929 cells, immortalized mouse testicular germ cells, 3T3, Vero, COS, HeLa, NT2, MG63, M24 and A375 metastatic melanoma, gliomas U251 and D54, HT1080 fibrosarcoma cells, and Sf9 insect cells.

It works extremely well on embryonic and neuronal stem cells; mono layers of stem cells can be grown after passaging with Accutase. It preserves most epitopes for subsequent flow cytometry analysis. Accutase does not need to be neutralized when passaging adherent cells. The addition of more media after the cells are split dilutes Accutase so it is no longer able to detach cells. It also does not need to be aliquoted. A bottle is stable in the refrigerator for 2 months.

Specifications

Specifications

  • Size:
    100 ml
  • Cat. No.:
    490007-741
  • Enzyme Name:
    ICT Accutase Cell Detachment Solution

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.