EpiXtract® Total Histone Extraction Kit (High Throughput), Enzo Life Sciences
10135-960
:
- Description:EpiXtract® Total Histone Extraction Kit (High Throughput)
- Cat. No.:10135-960
- No. of reactions:96
- Supplier no.:45013-0002
Rapid, reliable, high throughput method to isolate histone proteins in mammalian cells for histone modification applications.
For the extraction of total histone proteins from mammalian cells in a 96 well plate format.
EpiXtract® Total Histone Extraction Kit (High Throughput) provides a simple and reliable method to isolate histone proteins in 1 hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
: Contains Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .