BioArray™ Methylated DNA IP Kit, Enzo Life Sciences
Catalog # 10136-010
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Method / Format:Affinity resin
- Prep size:Scalable
- Refine target molecule:Methylated DNA
- Sample type:DNA preps
- Target molecule:DNA
- Description:BioArray™ Methylated DNA IP Kit
- Size:10 Tests
- Description:BioArray™ Methylated DNA IP Kit
- Cat. No.:10136-010
- Supplier no.:ENZ-45012-0010
Specifications
About this item
Enrich for methylated DNA (100-fold) from sample for complete diverse downstream applications
The BioArray™ methylated DNA IP kit features IP technology for the enrichment of 5-methylcytosine-containing DNA from any pool of fragmented genomic DNA for use in genome-wide methylation analysis. The kit includes a highly specific anti-5-methylcytosine monoclonal antibody for the “capture” and separation of methylated DNA from non-methylated DNA in only a few hours.
Ordering information: Contains Protein A Beads, Anti-5 methylcytosine, Control DNA, Control primer 1 , Control primer 2, IP Buffer, DNA denaturing Buffer, Control mouse DNA (methylated/nonmethylated), DNA elution buffer, Rods (magnetic).