BBL™ Yeast Extract, Life Technologies
Catalog # 90000-442
Supplier: Avantor
CAS Number:
Specifications
- Description:Yeast Extract
- Size:454 g
- Cat. No.:90000-442
- Supplier no.:211929
Specifications
About this item
BBL™ Yeast Extract is a concentrate of the water soluble portion of autolyzed yeast.
- Animal-free
- Water-soluble portion of autolyzed yeast
- Provides vitamins, amino acids, peptides, and neucleosides
- Used in a variety of animal health vaccines
BBL™ Yeast Extract is a concentrate of the water soluble portion of autolyzed yeast. This animal-free Yeast Extract provides a unique profile of essential water soluble vitamins, amino acids, peptides, neucleosides with a lower carbohydrate level. Customers are currently using BBL™ Yeast Extract is in a variety of animal health vaccines.