ContactUsLinkComponent
HighDef® Yellow IHC Chromogen (HRP), Enzo Life Sciences
HighDef® Yellow IHC Chromogen (HRP), Enzo Life Sciences
Catalog # 10049-340
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
HighDef® Yellow IHC Chromogen (HRP), Enzo Life Sciences
Catalog # 10049-340
Supplier:  Enzo Life Sciences
Supplier Number:  950-170-0030
CAS Number:  
Specifications
  • Description:
    HighDef® Yellow IHC Chromogen (HRP)
  • Size:
    30 mL
  • Cat. No.:
    10049-340
  • Supplier no.:
    950-170-0030
Specifications
About this item
Unique high definition yellow IHC chromogen for HRP-activated development.
HighDef® yellow IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridization systems. With at least 1 day working stability, this substrate/chromogen system can be used in any automation system as well as manual use.
Ordering information: Contains HighDef® yellow IHC chromogen (HRP) substrate buffer and chromogen, Empty mixing bottle.