HighDef® Blue IHC Chromogen (AP), Enzo Life Sciences
10049-336
:
- Description:HighDef® Blue IHC Chromogen (AP)
- Size:30 mL
- Cat. No.:10049-336
- Supplier no.:950-150-0030
High definition IHC chromogen producing brilliant blue color for AP-activated development.
HighDef® blue IHC chromogen (AP) is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems.
: Contains HighDef® blue IHC chromogen (AP) substrate buffer and chromogen, Empty mixing bottle.