HighDef® Red IHC Chromogen (AP, Plus), Enzo Life Sciences
Catalog # 10049-334
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:HighDef® Red IHC Chromogen (AP Plus)
- Size:30 mL
- Cat. No.:10049-334
- Supplier no.:950-141-0030
Specifications
About this item
High definition IHC chromogen producing brilliant red color for AP-activated development.
HighDef® red IHC chromogen (AP, plus) is intended to be used in conjuction with alkaline phosphatase (AP)-based immunostaining or in situ hybridization systems that run on automated staining instruments as well as manual use.
Ordering information: Contains HighDef® red IHC chromogen (AP, plus) substrate buffer and chromogen, Empty mixing bottle.