About this item
Structurally and mechanistically different compounds for studying the roles of pro- and anti-autophagic molecules in cells and in vitro applications.
The Screen-well Autophagy library contains 94 compounds with defined autophagy-inducing or -inhibitory activity. Compounds are dissolved in DMSO at 10mM or 1mM and aliquoted into deep-well plates at 100 or 500 µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-autophagic molecules in cells as well as for use in in vitro applications.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Concentration:DMSO solutions (10mM) except for Bafilomycin A which is provided at 1mM in DMSO.